2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%

Danger

Synonyms: AlbalonNaphazoline Hydrochloride

AC17966-0250 AC17966-1000
200040-376EA 159 USD
200040-376 200034-056
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Formula: C₁₄H₁₅ClN₂
MW: 246.738 g/mol
Melting Pt: 254…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00012554
CAS Number: 550-99-2
EINECS: 208-989-2
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR