Tetrathiafulvalene 99+%

Synonyms: 2,2'-Bi(1,3-dithiolylidene)TTF

AC17941-0010 AC17941-2500
200014-786EA 181 USD
200014-786 103511-052
Tetrathiafulvalene 99+%
Tetrathiafulvalene
Formula: C₆H₄S₄
MW: 204.356 g/mol
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR