PTFE

Synonyms: TeflonPolytetrafluoroethylene

AC17893-1000 AC17893-0050 AC17893-2500
200044-528EA 38.88 USD
200044-528 103466-342 200032-656
PTFE
PTFE
Formula: (CF₂CF₂)n
Boiling Pt: >400 °C (760 mmHg)
Melting Pt: 327 °C
Density: 2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS Number: 9002-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR