Poly(phenylene sulfide), M.W. ∼ 10000, powder

AC17885-0050
103466-338EA 15 USD
103466-338
Poly(phenylene sulfide), M.W. ∼ 10000, powder
Poly(phenylene sulfide)
Formula: (-C₆H₄S-)n
Density: 1.36 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084445
CAS Number: 9016-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR