Poly(methyl methacrylate), M.W. ∼35000, beads

Synonyms: PMMAMethyl Methacrylate PolymerMethacrylic Acid Methyl Ester Polymer

AC17876-5000 AC17876-0010 AC17876-0250
200014-104EA 24.23 USD
200014-104 200014-102 200032-644
Poly(methyl methacrylate), M.W. ∼35000, beads
Poly(methyl methacrylate)

Appearance: White Beads

Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Melting Pt: 105 °C
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS Number: 9011-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR