Polyethylene oxide, M.W. 100000-200000

Synonyms: PEO

AC17858-0050 AC17858-5000 AC17858-2500
200032-638EA 25.63 USD
200032-638 200032-642 200020-430
Polyethylene oxide, M.W. 100000-200000
Polyethylene oxide
Formula: (-CH₂CH₂O-)n
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00147970
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR