Cellulose acetate ∼ M.W. 100,000

AC17778-5000 AC17778-0250
200011-574EA 95.3 USD
200011-574 200030-672
Cellulose acetate ∼ M.W. 100,000
Cellulose acetate

Appearance: White Powder

Formula: C₂H₄O₂·(C₆H₁₀O₅)n
Melting Pt: 240 °C
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00081496
CAS Number: 9004-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR