Dimethyl (2-oxopropyl)phosphonate 95%

Synonyms: Dimethyl acetylmethylphosphonate

AC17761-0250 AC17761-0050
200012-092EA 100 USD
200012-092 200006-310
Dimethyl (2-oxopropyl)phosphonate 95%
Dimethyl (2-oxopropyl)phosphonate
Formula: C₅H₁₁O₄P
MW: 166.12 g/mol
Boiling Pt: 76…79 °C (3 mmHg)
Flash Pt: 93 °C
Storage Temperature: Ambient
EINECS: 224-110-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR