1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%

Danger

Synonyms: 1,3-Di(aminomethyl)cyclohexaneHexahydro-m-xylylenediamineCyclohexane-1,3-diyldimethanamine

AC17748-0050 AC17748-5000 AC17748-1000
200027-624EA 80.6 USD
200027-624 200021-622 200040-030
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture)

Appearance: Clear colorless Liquid

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.94 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00001522
CAS Number: 2579-20-6
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR