5,6,7,8-Tetrahydro-2-naphthol 98%

Warning

AC17747-0500 AC17747-0100
200030-020EA 169 USD
200030-020 200024-018
5,6,7,8-Tetrahydro-2-naphthol 98%
5,6,7,8-Tetrahydro-2-naphthol
Formula: C₁₀H₁₂O
Melting Pt: 58…62 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001738
CAS Number: 1125-78-6
EINECS: 214-410-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR