Borane dimethylamine complex 98+%

Danger

Synonyms: Boron hydride dimethylamineBorane, dimethylamine complex

AC17731-0050
103524-116EA 32.4 USD
103524-116
Borane dimethylamine complex 98+%
Borane dimethylamine complex
Formula: C₂H₁₀BN
MW: 58.92 g/mol
Melting Pt: 33…37 °C
Flash Pt: 65 °C
Storage Temperature: Refrigerator
EINECS: 200-823-7
UN: 2926
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR