Ethyl-4-chloroacetoacetate 98%

Danger

Synonyms: Ethyl 4-chloro-3-oxobutanoate

AC17764-0010 AC17764-2500 AC17764-0500
200012-368EA 29.5 USD
200012-368 103526-346 200019-076
Ethyl-4-chloroacetoacetate 98%
Ethyl 4-chloroacetoacetate
Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 103 °C (16 mmHg)
Melting Pt: –8 °C
Density: 1.21 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000939
CAS Number: 638-07-3
EINECS: 211-317-0
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR