4-Methoxy-N-methylaniline 98%

Warning

Synonyms: N-Methyl-p-anisidine

AC17722-0010 AC17722-0100
200032-296EA 41.1 USD
200032-296 200026-218
4-Methoxy-N-methylaniline 98%
4-Methoxy-N-methylaniline
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 135…136 °C (19 mmHg)
Melting Pt: 33…36 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008399
CAS Number: 5961-59-1
EINECS: 227-735-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR