Iodosobenzene diacetate 98%

Synonyms: Iodobenzene diacetate(Diacetoxyiodo)benzene

AC17656-1000 AC17656-0250 AC17656-5000
200001-828EA 156 USD
200001-828 103526-334 200001-826
Iodosobenzene diacetate 98%
Iodosobenzene diacetate
Formula: C₁₀H₁₁IO₄
MW: 322.1 g/mol
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00008692
CAS Number: 3240-34-4
EINECS: 221-808-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR