Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water

Danger

AC17579-1000 AC17579-0025 AC17579-5000
102777-932EA 1492 USD
102777-932 200058-390 200058-388
Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water
Pyruvaldehyde (Methylglyoxal)
Formula: C₃H₄O₂
Boiling Pt: 72 °C
Melting Pt: –20 °C
Density: 1.2 g/cm³
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR