Sulfuric acid-[D₂] 98% (99.5% D) (w/w) in D₂O for NMR spectroscopy

Danger

Synonyms: Sulfuric acid-[²H₂]Sulphuric acid-[²H₂]

AC17548-0050 AC17548-0500 AC17548-1000
200058-578EA 393.46 USD
200058-578 200058-576 102773-978
Sulfuric acid-[D₂] 98% (99.5% D) (w/w) in D₂O for NMR spectroscopy
Sulfuric acid-[D₂]
Formula: D₂SO₄
MW: 100.09 g/mol
Density: 1.87 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003542
CAS Number: 13813-19-9
UN: 1830
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR