Triethyloxonium tetrafluoroborate 1 M in methylene chloride

Danger

AC17511-5000 AC17511-1000 AC17511-0250
200058-788EA 71.31 USD
200058-788 200058-790 103526-328
Triethyloxonium tetrafluoroborate 1 M in methylene chloride
Triethyloxonium tetrafluoroborate
Formula: C₆H₁₅BF₄O
Density: 1.328 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00044423
CAS Number: 368-39-8
EINECS: 206-705-1
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR