3-Phenylpropionitrile 99+%

Danger

Synonyms: Hydrocinnamonitrile

AC17399-0100
200019-342EA 47.3 USD
200019-342
3-Phenylpropionitrile 99+%
3-Phenylpropionitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Melting Pt: 2…1 °C
Density: 1 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001961
CAS Number: 645-59-0
EINECS: 211-447-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR