Methyl-α-D-glucopyranoside 99%

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triolα-D-Methylglucoside

AC17384-1000 AC17384-0025 AC17384-5000
200011-120EA 102 USD
200011-120 200011-118 103509-228
Methyl-α-D-glucopyranoside 99%
Methyl α-D-glucopyranoside
Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Boiling Pt: 200 °C (.2 mmHg)
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00064086
EINECS: 202-571-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR