(±)-Methyl-2-bromopropionate 99%

Danger

Synonyms: Methyl 2-bromopropanoate

AC17339-1000 AC17339-5000
200031-896EA 176 USD
200031-896 200025-866
(±)-Methyl-2-bromopropionate 99%
(±)-Methyl 2-bromopropionate
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 143…145 °C (19 mmHg)
Density: 1.497 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000143
CAS Number: 5445-17-0
EINECS: 226-642-3
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR