Ethyl-4-bromobutyrate 97+%

Warning

Synonyms: Ethyl 4-bromobutyrate

AC17335-5000 AC17335-1000
200012-366EA 93.1 USD
200012-366 200025-302
Ethyl-4-bromobutyrate 97+%
Ethyl 4-bromobutyrate
Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 80…82 °C (10 mmHg)
Density: 1.36 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000259
CAS Number: 2969-81-5
EINECS: 221-005-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR