Abietic acid 85%

Warning

AC17313-0250 AC17313-1000
103526-310EA 77.4 USD
103526-310 200010-886
Abietic acid 85%
Abietic acid
Formula: C₂₀H₃₀O₂
Storage Temperature: Ambient
MDL Number: MFCD03423567
CAS Number: 514-10-3
EINECS: 208-178-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR