Sodium deuteroxide 30% in D₂O (99% D) (Isotopic) for NMR spectroscopy

Danger

AC17289-0200
200058-504EA 75.69 USD
200058-504
Sodium deuteroxide 30% in D₂O (99% D) (Isotopic) for NMR spectroscopy
Sodium deuteroxide
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037669
EINECS: 237-825-2
UN: 1824
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR