Ethyl malonyl chloride 90% tech.

Danger

Synonyms: Ethyl Malonyl Chloride

AC17269-1000 AC17269-0250 AC17269-0050
200006-494EA 161 USD
200006-494 200012-416 103526-306
Ethyl malonyl chloride 90% tech.
Ethyl malonyl chloride
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 79…80 °C (25 mmHg)
Density: 1.17 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000736
EINECS: 252-934-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR