EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) 99% for biochemistry
Synonyms:
HEPPS
, 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid
, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulfonic acid
, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid
, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid
, 4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acid
, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid)
, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid)
, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulfonic acid
, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid
, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid
Formula:
C₉H₂₀N₂O₄S MW: 252.33 g/mol Melting Pt: 237…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006160 CAS Number: 16052-06-5 EINECS: 240-198-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...