Chloroacetone 96% stabilized

Danger

AC17347-0250 AC17347-0010 AC17347-0050 AC17347-2500
102775-318EA 98.8 USD
102775-318 102774-162 102773-692 102777-092
Chloroacetone 96% stabilized
Chloroacetone
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –44.5 °C
Density: 1.16 g/cm³
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS Number: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR