Diatomaceous earth, Celite®, high purity, analytical grade for analysis analytical grade
|
Warning
|
Synonyms:
Celite®
, Diatomite
, Infusorial earth
, Diatomaceous silica
, Celite
, Kieselguhr
AC17346-0250
AC17346-0100
200024-692EA
87.34
USD
200024-692
103516-554
Diatomaceous earth, Celite®, high purity, analytical grade for analysis analytical grade
Diatomaceous earth
Formula:
SiO₂ MW: 60.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00132803 CAS Number: 61790-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...