4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical

Danger

Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl

AC17273-0050 AC17273-0010 AC17273-2500
102774-362EA 44.5 USD
102774-362 200017-336 200041-782
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR