Bis(dimethylamino)phosphoryl chloride 90% tech.
|
Danger
|
Synonyms:
Phosphoric Acid Bis(dimethylamido) Chloride
, N,N,N',N'-Tetramethylphosphorodiamidic Chloride
, Bis(dimethylamido)phosphoric chloride
, Bis(dimethylamino)phosphorochloridate
AC17272-0500
AC17272-0100
200024-576EA
54.4
USD
200024-576
102775-940
Bis(dimethylamino)phosphoryl chloride 90% tech.
Bis(dimethylamino)phosphoryl chloride
Appearance: Clear colorless to light yellow Liquid
Formula:
C₄H₁₂ClN₂OP MW: 170.58 g/mol Boiling Pt: 246…247 °C (760 mmHg) Density: 1.15 g/cm³ Flash Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008302 CAS Number: 1605-65-8 UN: 2922 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...