4-Chloro-7-nitrobenzofurazan 98%

Warning

Synonyms: NBD-ClNBD chloride4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole

AC17239-0010 AC17239-0250 AC17239-0050
103466-026EA 281 USD
103466-026 200007-118 200020-004
4-Chloro-7-nitrobenzofurazan 98%
4-Chloro-7-nitrobenzofurazan
Formula: C₆H₂ClN₃O₃
MW: 199.55 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005808
CAS Number: 10199-89-0
EINECS: 233-496-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR