2-Methyl-1-tetralone 98%
|
Warning
|
Synonyms:
2-Methyl-3,4-dihydronaphthalen-1(2H)-one
AC17237-0025
200016-660EA
164
USD
200016-660
2-Methyl-1-tetralone 98%
2-Methyl-1-tetralone
Formula:
C₁₁H₁₂O MW: 160.22 g/mol Boiling Pt: 127…131 °C (12 mmHg) Density: 1.06 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001690 CAS Number: 1590-08-5 EINECS: 216-461-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...