Dichloromethylenedimethyliminium chloride 98%

Danger

Synonyms: Phosgene-iminium ChlorideDichloromethylenedimethylammonium Chloride

AC17221-0050 AC17221-0250 AC17221-1000
200038-508EA 117 USD
200038-508 103531-670 200044-466
Dichloromethylenedimethyliminium chloride 98%
Dichloromethylenedimethyliminium chloride

Appearance: White to light yellow Powder or crystals

Formula: C₃H₆Cl₃N
MW: 162.44 g/mol
Melting Pt: 181…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011870
CAS Number: 33842-02-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR