N-Carbobenzoxy-L-aspartic acid 98%

Warning

Synonyms: N-Cbz-L-aspartic AcidN-Carbobenzyloxy-L-aspartic acidZ-Asp-OH

AC17205-0250 AC17205-1000
200032-184EA 60.1 USD
200032-184 103466-102
N-Carbobenzoxy-L-aspartic acid 98%
N-Carbobenzoxy-L-aspartic acid
Formula: C₁₂H₁₃NO₆
MW: 267.24 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00002719
CAS Number: 1152-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR