2,2-Diphenylglycine 98%
Synonyms:
2-Amino-2,2-diphenylacetic acid
, alpha,alpha-Diphenylglycine
, Diphenylglycin
AC17172-0050
AC17172-0250
AC17172-1000
103510-706EA
778
USD
103510-706
200018-068
200018-070
2,2-Diphenylglycine 98%
2,2-Diphenylglycine
Formula:
C₁₄H₁₃NO₂ MW: 227.26 g/mol Melting Pt: 244…247 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008048 CAS Number: 3060-50-2 EINECS: 221-299-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...