3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%

Warning

Synonyms: 3-(2-Pyridyl)-5,6-bis(4-sulphophenyl)-1,2,4-triazine disodium salt hydrate5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulphonic acid disodium salt hydrateFerroZine(TM) Iron reagent disodium salt hydrate3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine disodium salt hydratePDT disulfonate hydratePDT disulphonate hydrate5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate

AC17101-0010 AC17101-0050
200016-788EA 52.8 USD
200016-788 200009-614
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate
Formula: C₂₀H₁₂N₄Na₂O₆S₂·xH₂O
MW: 514.44 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00717621
CAS Number: 1264198-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR