1-Phenylcyclohexanecarboxylic acid 95%

Synonyms: 1-Phenyl-1-cyclohexanecarboxylic acid

AC17075-0500 AC17075-0100
200015-894EA 72.5 USD
200015-894 200027-946
1-Phenylcyclohexanecarboxylic acid 95%
1-Phenylcyclohexanecarboxylic acid
Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 119…124 °C
Storage Temperature: Ambient
EINECS: 214-495-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR