1-(4-Chlorophenyl)cyclobutanecarboxylic acid 94%
|
Warning
|
AC17072-0050
AC17072-1000
AC17072-0250
200015-422EA
192
USD
200015-422
103510-698
200015-418
1-(4-Chlorophenyl)cyclobutanecarboxylic acid 94%
1-(4-Chlorophenyl)cyclobutanecarboxylic acid
Formula:
C₁₁H₁₁ClO₂ MW: 210.6568 g/mol Melting Pt: 80…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001324 CAS Number: 50921-39-6 EINECS: 256-847-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...