1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 96%

Synonyms: 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid

AC17071-0500 AC17071-0100
200004-588EA 152 USD
200004-588 103510-696
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 96%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formula: C₁₁H₁₂O₃
MW: 192.2112 g/mol
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00019223
CAS Number: 16728-01-1
EINECS: 240-788-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR