(1-Ethoxyethylidene)malononitrile 99+%
|
Warning
|
Synonyms:
2-(1-Ethoxy-ethylidene)-malononitrile
, 2-(1-Ethoxyethylidene)malononitrile
AC17029-1000
AC17029-0250
200008-152EA
272
USD
200008-152
200039-252
(1-Ethoxyethylidene)malononitrile 99+%
(1-Ethoxyethylidene)malononitrile
Formula:
C₇H₈N₂O MW: 136.15 g/mol Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001852 CAS Number: 5417-82-3 EINECS: 226-521-5 UN: 3439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...