1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 99%

Synonyms: 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranoseβ-D-Ribofuranose 1-acetate 2,3,5-tribenzoate1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose

AC17011-0100
200018-290EA 91.1 USD
200018-290
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 99%
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
Formula: C₂₈H₂₄O₉
MW: 504.5 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005357
CAS Number: 6974-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR