Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-Glucose, anhydrous ≥99%

Supplier: ACROS ORGANICS MS

Synonyms: alpha-D(+)-Glucose

AC17008-0025 AC17008-0250 AC17008-0010
200057-396EA 42.83 USD InStock
200057-396 103516-540 200057-394
α-D-Glucose, anhydrous ≥99%
α-D-Glucose


Formula: C₆H₁₂O₆
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00063774
CAS Number: 492-62-6
EINECS: 207-757-8
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥99.0 % (ELSD)
Appearance of solution (10g in 15ml water) Passes test
Related substances ≤0.4 % (Maltose + Isomaltose)
Related substances ≤0.2 % (Maltotriose)
Related substances ≤0.15 % (Fructose)
Specific optical rotation +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/diluteNH3) (Anhydrous substance)
Soluble starch, sulfite ≤15 ppm
Dextrin Passes test


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty