Methyl (diethylphosphono)acetate 97%

Warning

Synonyms: Methyl 2-(diethoxyphosphoryl)acetateDiethyl methoxycarbonylmethanephosphonate

AC16998-0100
200013-288EA 53.6 USD
200013-288
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate
Formula: C₇H₁₅O₅P
MW: 210.17 g/mol
Boiling Pt: 127…131 °C (9 mmHg)
Density: 1.14 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009081
CAS Number: 1067-74-9
EINECS: 213-938-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR