Acetamidine hydrochloride 98+%
|
Warning
|
AC17021-5000
AC17021-1000
AC17021-0100
200010-898EA
23.4
USD
200010-898
200005-610
103510-688
Acetamidine hydrochloride 98+%
Acetamidine hydrochloride
Formula:
C₂H₇ClN₂ Melting Pt: 160…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013016 CAS Number: 124-42-5 EINECS: 204-700-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...