Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
|
Warning
|
Synonyms:
Dibenzo-18-crown-6
AC16980-0500
AC16980-0100
200037-024EA
128
USD
200037-024
200011-966
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
Formula:
C₂₀H₂₄O₆ MW: 360.41 g/mol Boiling Pt: 380…384 °C (679 mmHg) Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005098 CAS Number: 14187-32-7 EINECS: 238-041-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...