1,3-Dibromo-5,5-dimethylhydantoin 98%
|
Danger
|
Synonyms:
1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione
AC16954-5000
AC16954-0025
AC16954-0250
103531-636EA
226
USD
103531-636
200015-556
200008-462
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin
Formula:
C₅H₆Br₂N₂O₂ MW: 285.92 g/mol Melting Pt: 187…191 °C (dec.) Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003189 CAS Number: 77-48-5 EINECS: 201-030-9 UN: 3085 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...