Hydantoin 99%

Synonyms: Imidazolidine-2,4-dione

AC16870-1000 AC16870-5000
200037-616EA 145 USD
200037-616 200019-338
Hydantoin 99%
Hydantoin
Formula: C₃H₄N₂O₂
MW: 100.08 g/mol
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00005259
CAS Number: 461-72-3
EINECS: 207-313-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR