Tetrahydro-4H-thiopyran-4-one 99%
|
Warning
|
Synonyms:
Tetrahydrothiopyran-4-one
AC16798-0050
200020-874EA
123
USD
200020-874
Tetrahydro-4H-thiopyran-4-one 99%
Tetrahydro-4H-thiopyran-4-one
Formula:
C₅H₈OS MW: 116.18 g/mol Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006660 CAS Number: 1072-72-6 EINECS: 214-015-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...