Azidotrimethylsilane 94%

Danger

Synonyms: Trimethylsilyl azide

AC16793-2500
103526-216EA 671 USD
103526-216
Azidotrimethylsilane 94%
Azidotrimethylsilane
Formula: C₃H₉N₃Si
Boiling Pt: 95…96 °C (760 mmHg)
Density: 0.876 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001986
CAS Number: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR