3-(3,4-Dihydroxyphenyl)-L-alanine 99%

Warning

Synonyms: DoparL-dopa3-Hydroxy-L-tyrosineLevodopaL-β-3,4-Dihydroxyphenyl-Alanine

AC16753-5000 AC16753-1000 AC16753-0250 AC16753-0050
200006-808EA 186 USD
200006-808 103464-944 103464-946 200001-588
3-(3,4-Dihydroxyphenyl)-L-alanine 99%
3-(3,4-Dihydroxyphenyl)-L-alanine
Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002598
EINECS: 200-445-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR