N-Carbethoxy-4-piperidone 98.5+%
|
Warning
|
Synonyms:
N-Ethoxycarbonyl-4-piperidone
, 4-Piperidone-N-carboxylic acid ethyl ester
, Ethyl4-oxo-1-piperidinecarboxylate
AC16724-0100
AC16724-0500
200020-024EA
31.4
USD
200020-024
200020-026
N-Carbethoxy-4-piperidone 98.5+%
N-Carbethoxy-4-piperidone
Formula:
C₈H₁₃NO₃ MW: 171.19 g/mol Boiling Pt: 95…98 °C (1 mmHg) Density: 1.135 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006188 CAS Number: 29976-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...